missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethylenedimethyliminium Chloride 97.0+%, TCI America™
$76.43 - $229.29
Chemical Identifiers
CAS | 33842-02-3 |
---|---|
Molecular Formula | C3H6Cl3N |
Molecular Weight (g/mol) | 162.44 |
MDL Number | MFCD00011870 |
InChI Key | NRNFKRFWZQQDMD-UHFFFAOYSA-M |
Synonym | dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride |
PubChem CID | 2723945 |
IUPAC Name | dichloro(dimethylamino)methylium chloride |
SMILES | [Cl-].CN(C)[C+](Cl)Cl |
Chemical Identifiers
33842-02-3 | |
162.44 | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
2723945 | |
[Cl-].CN(C)[C+](Cl)Cl |
C3H6Cl3N | |
MFCD00011870 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
dichloro(dimethylamino)methylium chloride |
Specifications
33842-02-3 | |
C3H6Cl3N | |
5 g | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
[Cl-].CN(C)[C+](Cl)Cl | |
162.44 | |
162.44 | |
Crystalline Powder |
White-Yellow | |
MFCD00011870 | |
3261 | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
dichloro(dimethylamino)methylium chloride | |
2723945 | |
≥97.0% (T) | |
Dichloromethylenedimethyliminium Chloride |
Safety and Handling
TSCA : No