missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains amylene, >99.9%, Honeywell Burdick & Jackson
$3121.35 - $8983.83
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000508
|
Honeywell-Burdick and Jackson
CS29956 |
56 L | Returnable Steel Drum |
Each for $3,121.35
|
|
||||
6000503
|
Honeywell-Burdick and Jackson
CS299200 |
200 L | Returnable Steel Drum |
Each for $8,983.83
|
|
||||
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
75-09-2 | |
100.0000% | |
40°C | |
Returnable Steel Drum | |
MFCD00000881 | |
0.01% max. | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.94 | |
HPLC |
0.1 AU max. at 240 nm, 0.01 AU max. at 250 nm | |
1.33 g/cm3 | |
56 L | |
CH2Cl2 | |
UN1593 | |
1730800 | |
160% in Water at 20°C | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
>99.9% | |
Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains amylene, >99.9% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane