missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl phosphate, 98%
CAS: 1623-08-1 | C14H15O4P | 278.244 g/mol
Supplier: Thermo Scientific Chemicals L0881306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dibenzyl phosphate | |
78°C to 81°C | |
MFCD00004775 | |
2055755 | |
HDFFVHSMHLDSLO-UHFFFAOYSA-N | |
dibenzyl hydrogen phosphate | |
74189 | |
98% |
1623-08-1 | |
C14H15O4P | |
5 g | |
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883 | |
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2 | |
278.244 | |
278.24 |
Chemical Identifiers
1623-08-1 | |
278.244 | |
HDFFVHSMHLDSLO-UHFFFAOYSA-N | |
74189 | |
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2 |
C14H15O4P | |
MFCD00004775 | |
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883 | |
dibenzyl hydrogen phosphate |
Safety and Handling
EINECSNumber : 216-602-3
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only