missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberone 98.0+%, TCI America™

$54.06 - $147.25

Chemical Identifiers

CAS 1210-35-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003587
InChI Key BMVWCPGVLSILMU-UHFFFAOYSA-N
Synonym dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
PubChem CID 14589
IUPAC Name tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
SMILES O=C1C2=CC=CC=C2CCC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D180125G
View Documents
TCI America
D180125G
25 g
Each for $54.06
Only null left
 
D1801100G
View Documents
TCI America
D1801100G
100 g
Each for $147.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1210-35-1
208.26
BMVWCPGVLSILMU-UHFFFAOYSA-N
14589
O=C1C2=CC=CC=C2CCC2=CC=CC=C12
C15H12O
MFCD00003587
dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one

Specifications

1210-35-1
148°C
MFCD00003587
dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
O=C1C2=CC=CC=C2CCC2=CC=CC=C12
208.26
208.26
Crystalline Lumps
White-Yellow
C15H12O
25 g
BMVWCPGVLSILMU-UHFFFAOYSA-N
tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
14589
≥98.0% (GC)
Dibenzosuberone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2139

RTECSNumber : HP1149700

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'