missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl Hydrazodicarboxylate 98.0+%, TCI America™

$93.21 - $277.75

Chemical Identifiers

CAS 16466-61-8
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00015000
InChI Key TYSZETYVESRFNT-UHFFFAOYSA-N
Synonym di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
PubChem CID 85431
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
SMILES CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D42245G
View Documents
TCI America
D42245G
5 g
Each for $93.21
Only null left
 
D422425G
View Documents
TCI America
D422425G
25 g
Each for $277.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16466-61-8
232.28
TYSZETYVESRFNT-UHFFFAOYSA-N
85431
CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
C10H20N2O4
MFCD00015000
di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate

Specifications

16466-61-8
White
MFCD00015000
di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
232.28
232.28
Crystalline Powder
125°C
C10H20N2O4
5 g
TYSZETYVESRFNT-UHFFFAOYSA-N
tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
85431
≥98.0% (GC)
Di-tert-butyl Hydrazodicarboxylate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'