missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKZ Monoclonal Antibody (K49027_8F3), Invitrogen™
Mouse Monoclonal Antibody
$1557.73 - $2745.56
Specifications
Antigen | DGKZ |
---|---|
Clone | K49027_8F3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
Sequence of this protein is as follows: STGSKDVVRY LLDHAPPEIL DAVEENGETC LHQAAALGQR TICHYIVEAG ASLMKTDQQG DTPRQRAEKA QDTELAAYLE NRQHYQMIQR
DGKZ Monoclonal antibody specifically detects DGKZ in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
DGKZ | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide | |
104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z] | |
Recombinant human DGKZ (immunogen range is 832-921). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
DGKZ |
K49027_8F3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q13574, Q80UP3 | |
104418, 8525 | |
IgG1 | |
Protein A | |
Antibody |