missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Monohydrate, Powder, Spectrum™ Chemical
$439.35 - $4245.94
Chemical Identifiers
CAS | 77938-63-7 |
---|---|
Molecular Formula | C6H14O7 |
Molecular Weight (g/mol) | 198.17 |
InChI Key | SPFMQWBKVUQXJV-UHFFFAOYNA-N |
IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal hydrate |
SMILES | O.OCC(O)C(O)C(O)C(O)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610149
|
Spectrum Chemical Mfg Cor
D1018500GM |
500 g | Poly Bottle |
Each for $439.35
|
|
||||
18610150
|
Spectrum Chemical Mfg Cor
D101825KG |
2.5 kg | Poly Bottle |
Each for $1,365.03
|
|
||||
18610151
|
Spectrum Chemical Mfg Cor
D101845KGBL |
45 kg | Fiber Drum |
Each for $4,245.94
|
|
||||
Chemical Identifiers
77938-63-7 | |
198.17 | |
2,3,4,5,6-pentahydroxyhexanal hydrate |
C6H14O7 | |
SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
O.OCC(O)C(O)C(O)C(O)C=O |
Specifications
77938-63-7 | |
Poly Bottle | |
500 g | |
O.OCC(O)C(O)C(O)C(O)C=O | |
198.17 | |
5e-05 |
1 | |
C6H14O7 | |
SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
2,3,4,5,6-pentahydroxyhexanal hydrate | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.