missing translation for 'onlineSavingsMsg'
Learn More

Dextran 150,000, Powder, Spectrum™ Chemical

$280.63 - $32036.87

Chemical Identifiers

CAS 9004-54-0
Molecular Formula (C6H9O5)n(C6H10O5)m
MDL Number MFCD00130935
IUPAC Name Dextran
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610141
View Documents
Spectrum Chemical Mfg Cor
D101325GM
25 g Poly Bottle
Each for $280.63
Only null left
 
18610140
View Documents
Spectrum Chemical Mfg Cor
D1013100GM
100 g Poly Bottle
Each for $667.03
Only null left
 
18610139
View Documents
Spectrum Chemical Mfg Cor
D10131KG
1 kg Poly Pail
Each for $3,474.43
Only null left
 
18610142
View Documents
Spectrum Chemical Mfg Cor
D101325KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

9004-54-0
MFCD00130935
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
(C6H9O5)n(C6H10O5)m
Dextran

Specifications

White to Slightly Off White
9004-54-0
(C6H9O5)n(C6H10O5)m
25 g
Dextran
0.1
4.0 to 7.0
1
MFCD00130935
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
Reagent
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.