Learn More
3M™ DBI-SALA™ Ultra-Lok™ Self Retracting Lifeline- Cable
Stainless steel components, an anti-ratcheting brake system and a semi-sealed design provide the user with a system built to last. Lifeline will extend as the user moves away and retract automatically enabling the user to move about within a recommended working area at normal speeds.
Supplier:  3M™ 3504430C
 View more versions of this product
Catalog No. 29-632-338
Only null left
Explore available promotions
Description
Description
  • Rugged and extremely lightweight thermoplastic housing
  • Semi-sealed design
  • 50 ft. (15.2 m) galvanized steel cable lifeline
  • Stainless steel dynamic components
  • Internal energy absorption system
  • Anti-ratcheting twin disk brake system
  • Constant moving brake system pawls
  • Swiveling anchorage point
  • Swiveling snap hook with impact indicator
  • Reserve lifeline system
  • Ergonomic cable handle equipped with i-Safe™
Specifications
Specifications
Block to anchor point mounting type
Self-retracting Lifeline - Cable
Urethane Thermoplatic, Zinc-plated Steel, Galvanized Cable
Galvanized steel wire rope with swivel snap hook
30 ft.
141 kg
ANSI Z359.14, ANSI A10.32, ANSI Z359.1, OSHA 1926.502, OSHA 1910.66
9.14 m
141 kg
Construction, General Industrial, Transportation, Utilities, Wind Energy
310 lb.
3/16 in.
11 lb.
5 mm
310 lb.
Product Suggestions
Product Suggestions