Learn More
3M™ DBI-Sala™ Crossover Full-Body Harnesses
Lightweight polyester for all-day wear
Supplier:  3M™ 1103375C
 View more versions of this product
Catalog No. 19-809-414
Only null left
Explore available promotions
Description
Description
  • Feature patented crossover design with adjustable front D-ring, used for positioning or ladder climbing
  • Back D-ring is used for fall arrest or restraint
  • Friction buckles on shoulders and mating pass-through adjuster buckles on lower shoulder straps and leg straps
  • Side D-rings for additional restraint or positioning options
  • Suitable for fall arrest, restraint, or positioning
Compliance
Meet ANSI Z359.1-1992, ANSI A10.14-1991, and OSHA requirements.
Specifications
Specifications
Universal
Lightweight Polyester
Front, Side
Product Suggestions
Product Suggestions