Learn More
Applied Biosystems™ dAbz, 0.2 μmol ABI 3900 HT Columns
The 0.2μmol d(Abz) ABI 3900 HT columns are fully assembled, low-cost columns that produce high-quality oligonucleotides. These supports are filled with polystyrene, a material which has several advantages over the more conventional, controlled pore glass (CPG) filled columns.
Supplier: Applied Biosystems™ 4316675
Description
Designed for PerformanceThe greater hydrophobicity of polystyrene results in a higher yield of full-length product. Side reactions, such as extraneous chain growth, are minimized by the inert nature of this support surface.- Proprietary design and manufacturing methods ensure consistency and high quality to give you optimal synthesis quality.
- Hydrophobic, polystyrene-filled columns sustain an anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis.
- Polystyrene support allows synthesis of oligonucleotides of a variety of lengths, giving you flexibility in high-throughput oligonucleotide synthesis.
Specifications
Shipped at Room Temperature | |
200 Columns |
For Research Use Only. Not for use in diagnostic procedures.