missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Threonine, 99%
CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol
$74.67 - $750.26
Chemical Identifiers
CAS | 632-20-2 |
---|---|
Molecular Formula | C4H9NO3 |
Molecular Weight (g/mol) | 119.12 |
MDL Number | MFCD00064269 |
InChI Key | AYFVYJQAPQTCCC-STHAYSLISA-N |
Synonym | d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth |
PubChem CID | 69435 |
ChEBI | CHEBI:16398 |
IUPAC Name | (2R,3S)-2-amino-3-hydroxybutanoic acid |
SMILES | C[C@H](O)[C@@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2117706
|
Thermo Scientific Chemicals
B2117706 |
5 g |
Each for $74.67
|
|
|||||
AAB2117714
|
Thermo Scientific Chemicals
B2117714 |
25 g |
Each for $227.73
|
|
|||||
AAB2117722
|
Thermo Scientific Chemicals
B2117722 |
100 g |
Each for $750.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
632-20-2 | |
119.12 | |
AYFVYJQAPQTCCC-STHAYSLISA-N | |
69435 | |
(2R,3S)-2-amino-3-hydroxybutanoic acid |
C4H9NO3 | |
MFCD00064269 | |
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth | |
CHEBI:16398 | |
C[C@H](O)[C@@H](N)C(O)=O |
Specifications
∼274°C (decomposition) | |
C4H9NO3 | |
1721643 | |
AYFVYJQAPQTCCC-STHAYSLISA-N | |
(2R,3S)-2-amino-3-hydroxybutanoic acid | |
119.12 | |
CHEBI:16398 | |
119.12 | |
D-Threonine |
632-20-2 | |
MFCD00064269 | |
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth | |
C[C@H](O)[C@@H](N)C(O)=O | |
5 g | |
69435 | |
+29° (c=5 in Water) | |
99% |
Safety and Handling
EINECSNumber : 211-171-8
RTECSNumber : XO8580000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only