missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol, 99.2%, Cell culture reagent, For HPLC analysis, MP Biomedicals™

$92.26 - $92.26

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00077932
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19468850
View Documents
MP Biomedicals Inc
0219468850
50 g
Each for $92.26
Only null left
 
Description

Description

Specifications

Chemical Identifiers

87-89-8
180.156
CDAISMWEOUEBRE-UHFFFAOYSA-N
892
cyclohexane-1,2,3,4,5,6-hexol
C6H12O6
MFCD00077932
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
CHEBI:24848
C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

87-89-8
6.2g/mL
MFCD00077932
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
C1(C(C(C(C(C1O)O)O)O)O)O
180.156
CHEBI:24848
Cell Culture Reagent
D-myo-inositol, Molecular biology and Cell biology
White
C6H12O6
50 g
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
892
180.2
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-781-2

Recommended Storage : Store at room temperature.

SDS
missing translation for 'documents'

missing translation for 'documents'