missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Methionine, 99%
CAS: 348-67-4 | C5H11NO2S | 149.21 g/mol
$62.84 - $649.63
Chemical Identifiers
CAS | 348-67-4 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.21 |
MDL Number | MFCD00002622 |
InChI Key | FFEARJCKVFRZRR-SCSAIBSYSA-N |
Synonym | d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid |
PubChem CID | 84815 |
ChEBI | CHEBI:16867 |
IUPAC Name | (2R)-2-amino-4-methylsulfanylbutanoic acid |
SMILES | CSCC[C@@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2121306
|
Thermo Scientific Chemicals
B2121306 |
5 g |
Each for $62.84
|
|
|||||
AAB2121314
|
Thermo Scientific Chemicals
B2121314 |
25 g |
Each for $220.66
|
|
|||||
AAB2121322
|
Thermo Scientific Chemicals
B2121322 |
100 g |
Each for $649.63
|
|
|||||
Description
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.
Solubility
Soluble in water. 53 g/L (20°C).
Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
Chemical Identifiers
348-67-4 | |
149.21 | |
FFEARJCKVFRZRR-SCSAIBSYSA-N | |
84815 | |
(2R)-2-amino-4-methylsulfanylbutanoic acid |
C5H11NO2S | |
MFCD00002622 | |
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
CHEBI:16867 | |
CSCC[C@@H](N)C(O)=O |
Specifications
∼280°C (decomposition) | |
C5H11NO2S | |
1722293 | |
d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
FFEARJCKVFRZRR-SCSAIBSYSA-N | |
(2R)-2-amino-4-methylsulfanylbutanoic acid | |
149.21 | |
CHEBI:16867 | |
149.21 | |
D-Methionine |
348-67-4 | |
MFCD00002622 | |
14,5975 | |
Soluble in water. 53g/L (20°C). | |
CSCC[C@@H](N)C(O)=O | |
5 g | |
84815 | |
−23° (c=2 in 2N HCl) | |
99% |
Safety and Handling
H500
EINECSNumber : 206-483-6
RTECSNumber : PD0455000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only