missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, MP Biomedicals™

D-Mannitol, ACS

$410.74 - $548.23

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym Mannite
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP218379090
View Documents
MP Biomedicals Inc
0218379090
500 g
Each for $548.23
Only null left
 
MP218379091
View Documents
MP Biomedicals Inc
0218379091
1 kg
Each for $410.74
Only null left
 
Description

Description

  • Used in sweetness inhibition studies.
  • A sugar alcohol sweet tastant.
Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064287
Mannite
OCC(O)C(O)C(O)C(O)CO

Specifications

69-65-8
C6H14O6
500 g
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
ACS
167°C to 170°C (literature)
MFCD00064287
Mannite
OCC(O)C(O)C(O)C(O)CO
182.17
Videos
Safety and Handling

Safety and Handling

storageNote1 : Store at Room Temperature(15-30°C).

SDS
missing translation for 'documents'

missing translation for 'documents'