missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Mandelic Acid 99.0+%, TCI America™

$121.16 - $1019.63

Chemical Identifiers

CAS 611-71-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064251
InChI Key IWYDHOAUDWTVEP-SSDOTTSWSA-N
Synonym r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
PubChem CID 11914
ChEBI CHEBI:17656
IUPAC Name (2R)-2-hydroxy-2-phenylacetic acid
SMILES O[C@@H](C(O)=O)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M066225G
View Documents
TCI America
M066225G
25 g
Each for $121.16
Only null left
 
M0662100G
View Documents
TCI America
M0662100G
100 g
Each for $339.25
Only null left
 
M0662500G
View Documents
TCI America
M0662500G
500 g
Each for $1,019.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

611-71-2
152.15
IWYDHOAUDWTVEP-SSDOTTSWSA-N
11914
(2R)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00064251
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
CHEBI:17656
O[C@@H](C(O)=O)C1=CC=CC=C1

Specifications

134°C
White to Yellow
MFCD00064251
IWYDHOAUDWTVEP-SSDOTTSWSA-N
(2R)-2-hydroxy-2-phenylacetic acid
11914
152.15
25 g
D-(-)-Mandelic Acid
611-71-2
C8H8O3
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
O[C@@H](C(O)=O)C1=CC=CC=C1
152.15
CHEBI:17656
≥99.0% (GC,T)
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1764

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'