missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glucurono-3,6-lactone, Spectrum™ Chemical
$196.17 - $22575.27
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610165
|
Spectrum Chemical Mfg Cor
G216025GM |
25 g | Amber Glass Bottle |
Each for $196.17
|
|
||||
18610164
|
Spectrum Chemical Mfg Cor
G2160100GM |
100 g | Amber Glass Bottle |
Each for $334.37
|
|
||||
18610168
|
Spectrum Chemical Mfg Cor
G2160500GM |
500 g | Amber Glass Bottle |
Each for $1,226.70
|
|
||||
18610167
|
Spectrum Chemical Mfg Cor
G21605KGBL |
5 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18610166
|
Spectrum Chemical Mfg Cor
G216025KGBL |
25 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
32449-92-6 |
Specifications
32449-92-6 | |
Amber Glass Bottle | |
Reagent |
1 | |
25 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.