Learn More
Thermo Scientific Chemicals D-(+)-Cellotriose, 95%
$301.86 - $877.36
Chemical Identifiers
CAS | 33404-34-1 |
---|---|
Molecular Formula | C18H32O16 |
Molecular Weight (g/mol) | 504.44 |
MDL Number | MFCD00069842 |
InChI Key | FYGDTMLNYKFZSV-ZWSAEMDYSA-N |
Synonym | cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide |
PubChem CID | 5287993 |
ChEBI | CHEBI:3528 |
IUPAC Name | (2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67702LB0
|
Thermo Scientific Chemicals
J67702LB0 |
5 mg |
Each for $301.86
|
|
|||||
AAJ67702MA
|
Thermo Scientific Chemicals
J67702MA |
10 mg |
Each for $441.33
|
|
|||||
AAJ67702MB
|
Thermo Scientific Chemicals
J67702MB |
25 mg |
Each for $877.36
|
|
|||||
Description
A 3-glucose cellulodextrin derived from cellulose degradation. An energy source for cellulosic bacteria. Can be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases. Also useful in saccharification and ethanol fermentation research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33404-34-1 | |
504.44 | |
FYGDTMLNYKFZSV-ZWSAEMDYSA-N | |
5287993 | |
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
C18H32O16 | |
MFCD00069842 | |
cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide | |
CHEBI:3528 | |
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
33404-34-1 | |
C18H32O16 | |
5 mg | |
Hygroscopic | |
Soluble in water at 50mg/ml /n | |
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O | |
504.44 | |
CHEBI:3528 | |
95% | |
D-(+)-Cellotriose |
White | |
MFCD00069842 | |
100353 | |
cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide | |
FYGDTMLNYKFZSV-ZWSAEMDYSA-N | |
(2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol | |
5287993 | |
504.44 | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only