missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Asparagine monohydrate
$103.08 - $291.23
Chemical Identifiers
CAS | 5794-24-1 |
---|---|
Molecular Weight (g/mol) | 150.13 |
InChI Key | RBMGJIZCEWRQES-UHFFFAOYNA-N |
IUPAC Name | 2-amino-3-carbamoylpropanoic acid hydrate |
SMILES | O.NC(CC(N)=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC204270010
|
Thermo Scientific Chemicals
204270010 |
1 g |
Each for $103.08
|
|
|||||
AC204270050
|
Thermo Scientific Chemicals
204270050 |
5 g |
Each for $144.50
|
|
|||||
AC204270250
|
Thermo Scientific Chemicals
204270250 |
25 g |
Each for $291.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5794-24-1 | |
RBMGJIZCEWRQES-UHFFFAOYNA-N | |
O.NC(CC(N)=O)C(O)=O |
150.13 | |
2-amino-3-carbamoylpropanoic acid hydrate |
Specifications
280.0°C | |
C4H10N2O4 | |
04, 471 | |
Solubility aq. soln.: 35 g/L (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene | |
O.NC(CC(N)=O)C(O)=O | |
1 g | |
150.13 |
5794-24-1 | |
H2NCOCH2CH(NH2)CO2H·H2O | |
14, 837 | |
RBMGJIZCEWRQES-UHFFFAOYNA-N | |
2-amino-3-carbamoylpropanoic acid hydrate | |
150.13 | |
D-Asparagine monohydrate |
RUO – Research Use Only