missing translation for 'onlineSavingsMsg'
Learn More

CytoScint ES, MP Biomedicals™

Liquid Scintillation Cocktails; CytoScint ES; MP Biomedical; 4 x 1 gal.; Biodegradable

$840.00 - $1532.86

Specifications

Chemical Name or Material Liquid Scintillation Cocktails
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
88245301
View Documents
MP Biomedicals Inc
0188245301
1 gal.
Each for $840.00
Only null left
 
88245304
View Documents
MP Biomedicals Inc
0188245304
4 x 1 gal.
Each for $1,532.86
Only null left
 
Description

Description

Specifications

Specifications

Liquid Scintillation Cocktails
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'