missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl bromide, 99%

CAS: 4333-56-6 | C3H5Br | 120.98 g/mol

$352.88 - $352.88

Chemical Identifiers

CAS 4333-56-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.98
MDL Number MFCD00001271
InChI Key LKXYJYDRLBPHRS-UHFFFAOYSA-N
Synonym cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
PubChem CID 78037
IUPAC Name bromocyclopropane
SMILES C1CC1Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111640250
SDS View Documents
Thermo Scientific Chemicals
111640250
25 mL Glass bottle
Each for $352.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4333-56-6
120.98
LKXYJYDRLBPHRS-UHFFFAOYSA-N
78037
C1CC1Br
C3H5Br
MFCD00001271
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
bromocyclopropane

Specifications

4333-56-6
1.5100g/mL
Authentic
Glass bottle
1.4590 to 1.461
MFCD00001271
1.51
Solubility in water: immiscible
C1CC1Br
120.98
120.98
Liquid
Colorless
-7°C
98.5% min. (GC)
C3H5Br
C3H5Br
25 mL
cyclopropyl bromide, cyclopropane, bromo, cyclopropylbromide, bromo-cyclopropan, bromo-cyclopropane, cyclopropylbrornide, bromanylcyclopropane, pubchem14417, bromocyclopropane, acmc-1aii5
LKXYJYDRLBPHRS-UHFFFAOYSA-N
bromocyclopropane
78037
99%
Cyclopropyl bromide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-375-7

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only