missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, Reagents

Cyclohexanone, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$101.47 - $495.97

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym Pimelic Ketone, Ketohexamethylene
IUPAC Name cyclohexanone
SMILES O=C1CCCCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-297
View Documents
Labchem Inc
C2193600100C
100 mL
Each for $101.47
Only null left
 
01-337-296
View Documents
Labchem Inc
C21936001C
1 L
Each for $495.97
Only null left
 
01-337-295
View Documents
Labchem Inc
C2193600500C
500 mL
Each for $256.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

108-94-1
98.15
Pimelic Ketone, Ketohexamethylene
O=C1CCCCC1
C6H10O
JHIVVAPYMSGYDF-UHFFFAOYSA-N
cyclohexanone

Specifications

-28.9°C
Colorless or Pale Yellow
100 mL
Liquid
C6H10O
JHIVVAPYMSGYDF-UHFFFAOYSA-N
cyclohexanone
Reagent
108-94-1
154°C
Glass Bottle
0.9478
Pimelic Ketone, Ketohexamethylene
O=C1CCCCC1
98.15
Cyclohexanone
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'