missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
$207.75 - $3221.85
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000479
|
Honeywell-Chromasolv
344961L |
1 L |
Each for $207.75
|
|
|||||
6000480
|
Honeywell-Chromasolv
3449625L |
2.5 L |
Each for $461.98
|
|
|||||
60046353
|
Honeywell-Chromasolv
344964X25L |
4 x 2.5 L |
Case of 4 Each for $1,594.58
|
|
|||||
6000481
|
Honeywell-Chromasolv
344964X4L |
4 x 4 L |
Case of 4 Each for $3,221.85
|
|
|||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Specifications
110-82-7 | |
1 L | |
−18°C (−.4°F) | |
C6H12 | |
168.8 mmHg (37.7°C), 77 mmHg (20°C) | |
1900225 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.16g/mol | |
CHROMASOLV™ |
0.779g/mL at 25°C | |
0.09 | |
500°F | |
MFCD00003814 | |
UN1145 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
≥99.5% (GC) | |
Cyclohexane |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
RTECSNumber : GU6300000