missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.0% min., ACS, MilliporeSigma™

$315.15 - $8132.25

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MCX22901
View Documents
Technidata
CX22901
1 L
Each for $315.15
Only null left
 
MCX22903
View Documents
Technidata
CX22903
4 L
Each for $460.59
Only null left
 
MCX229030
View Documents
Technidata
CX229030
200 L
Each for $8,132.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1

Specifications

110-82-7
Liquid
C6H12
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
Cyclohexane, For ACS analysis
1 L
99.0%, min.
-CH2(CH2)4CH2-
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.16
CHEBI:29005
ACS/NF/USP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'