missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves

FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves feature soft and flexible TPR pads that provide an additional layer of protection against impacts to the back of the hand and fingers. Nitrile coating on the palm improves grip capabilities when handling wet and oily parts.

$249.92 - $499.86

Specifications

ANSI Abrasion Rating ANSI 4
Applications Industrial
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A4
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17200350
View Documents
Wells Lamont™
I2469S
Small
Pack of 6 Pair for $249.92
Only null left
 
17200351
View Documents
Wells Lamont™
I2469M
Medium
Pack of 6 Pair for $249.92
Only null left
 
17200352
View Documents
Wells Lamont™
I2469L
Large
Pack of 6 Pair for $249.92
Only null left
 
17200353
View Documents
Wells Lamont™
I2469XL
X-Large
Dozen of 12 for $499.86
Case of 6 Dozen for $2,726.58
Only null left
 
17200354
View Documents
Wells Lamont™
I2469XXL
2X-Large
Dozen of 12 for $499.86
Case of 6 Dozen for $2,726.58
Only null left
 
Description

Description

  • A blend of high performance and cooling fibers extends comfort in warmer work environments
  • Soft and flexible TPR pads provide an additional layer of protection against blows to the back of the hand and fingers
  • Reinforced thumb crotch for extra protection between the thumb and forefinger
  • An ANSI Cut Level A4 ensures a high level of cut protection against sharp objects
  • Padded, sandy nitrile coated palm increases protection and improves grip
  • Ideal for handling parts in wet and oily environments
Specifications

Specifications

ANSI 4
Nitrile
ANSI A4
Specific
Anti-impact Gloves
ANSI/ISEA 105-16 Level 4
I2469
15
Industrial
Hi-vis Orange
Fully Textured
HPPE
Knit
Cuts, Abrasions, Impacts, Punctures
4
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'