missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ Cushion Trax™ Anti-Fatigue Mat
Cushion Trax™ is an anti-fatigue floor mat with a heavy-duty vinyl diamond-plate design and a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.
Supplier: NoTrax™ 479S0312GY
Description
- Cushion Trax™ is engineered to meet the toughest requirements of today's industrial applications
- It is particularly suited for multi-shift operations in dry work areas where the worker is standing for prolonged periods of time such as workstations and assembly lines
- The heavy-duty vinyl diamond-plate design provides non-directional traction, and is easy to clean
- The top surface is combined with a dense closed cell foam base utilizing NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
- NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate the slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
- The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat
Specifications
12 x 3 ft. | |
Smoke Gray | |
Manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual workstations. Suitable for commercial environments including hotels, reception desks, shop counters, department stores, banks, restaurants, etc. | |
14.2875 mm | |
9/16 in. | |
Certified by the NFSI (National Floor Safety Institute) |
12 ft. | |
3 ft. | |
Anti-fatigue Floor Mat | |
Nitricell, PVC | |
0.9144 m | |
3.6576 m |