missing translation for 'onlineSavingsMsg'
Learn More

Curcumin, Spectrum™ Chemical

$248.18 - $65116.15

Chemical Identifiers

CAS 458-37-7
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610080
View Documents
Spectrum Chemical Mfg Cor
C145210GM
10 g Amber Glass Bottle
Each for $248.18
Only null left
 
18610082
View Documents
Spectrum Chemical Mfg Cor
C145225GM
25 g Amber Glass Bottle
Each for $450.47
Only null left
 
18610081
View Documents
Spectrum Chemical Mfg Cor
C1452100GM
100 g Amber Glass Bottle
Each for $1,517.09
Only null left
 
18610084
View Documents
Spectrum Chemical Mfg Cor
C1452500GMBL
500 g Amber Glass Bottle
Each for $3,416.03
Only null left
 
18610083
View Documents
Spectrum Chemical Mfg Cor
C145225KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

458-37-7

Specifications

458-37-7
Amber Glass Bottle
Reagent
1
10 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.