missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Balance

The Cubis™ II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a range of accessories allows the individual adaptation to all weighing tasks.

$25032.52 - $45220.43

Specifications

Display Type Color Touchscreen
Type Balance
Legal for Trade Class I
View More Specs

Products 5
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
Catalog Number Mfr. No. Capacity Model Readability Price Quantity  
01-672-091
View Documents
Sartorius
MCA125SI3MTS
120 g MCA with Draft Shield D 0.01 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-672-090
View Documents
Sartorius
MCA225SI3MTS
220 g MCA with Draft Shield D 0.01 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-672-089
View Documents
Sartorius
MCA226SI3MTS
220 g MCA with Draft Shield D 0.005 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-672-088
View Documents
Sartorius
MCE125SI3MTS
120 g MCE with Draft Shield D 0.01 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-672-087
View Documents
Sartorius
MCE225SI3MTS
220 g MCE with Draft Shield D 0.01 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Leading performance. Monolithical weighing system, integrated climate sensors, individual sample holders Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Full end-to-end data integrity. 21 CFR Part 11 compli-ance, integrated audit-trail, state-of-the-art user management
  • Outstanding service support. Integrated status center, integrated service functions preventive maintenance on accredited standards
Specifications

Specifications

Color Touchscreen
Balance
Class I
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'