Learn More
Sartorius Cubis™ II 36P High-Capacity Essential Micro Balance
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
$48023.30 - $49448.90
Specifications
Connection Type USB A, USB B, USB C
Capacity 32 g, 10.1 g
Display Type Color TFT Touchscreen
Readability 0.01 mg, 0.001 mg
Material Stainless Steel
View More Specs
Products 4
Catalog Number Mfr. No. Model Price Quantity  
Catalog Number Mfr. No. Model Price Quantity  
01-675-447
View Documents
Sartorius
MCE36P3S00D
Standard Configuration with Draft Shield N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-675-444
View Documents
Sartorius
MCE36P3S00DMDS
MDS Configuration with Draft Shield N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-675-445
View Documents
Sartorius
MCE36P3S00DION
ION Configuration with Draft Shield N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-675-446
View Documents
Sartorius
MCE36P3S00DHWL
HWL Configuration with Draft Shield N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Ultra-high resolution with up to 61 million weighing steps (digits)
  • End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
  • Cleanability, part of compliance
  • Error-free operation. Individual QApp workflows, motorized auto-leveling
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer
  • Optional inner draft shield for best weighing performance
  • Housing: Stainless steel 1.4401, 1.4404, Aluminum, Plastic PBT, PA, Float glass Optiwhite, Control Unit: Aluminum, painted, Plastic PBT, PP, Float glass
Specifications
Specifications
USB A, USB B, USB C
Color TFT Touchscreen
Stainless Steel
Micro Balance
No
32 g, 10.1 g
0.01 mg, 0.001 mg
3.5 sec., 2.5 sec.
100/240 V
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'