Learn More
Tempshield™ Cryo-Gloves™ Blue Waterproof Wrist-Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
$554.57 - $585.60
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Wrist Length |
Color | Blue |
Hand | Specific |
Product Type | Waterproof Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
11-394-322
![]() |
Tempshield™
WRSWP |
Small |
Pair for $554.57
Case of 10 Pair for $5,041.60
|
|
|||||
11-394-323
![]() |
Tempshield™
WRMWP |
Medium |
Pair for $585.60
Case of 10 Pair for $5,323.70
|
|
|||||
11-394-324
![]() |
Tempshield™
WRLWP |
Large |
Pair for $585.60
Case of 10 Pair for $5,323.70
|
|
|||||
Description
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Waterproof Cryogenic Gloves | |
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes | |
-320°F | |
11 to 13 in. |
Wrist Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
28 to 33 cm |