Learn More
Applied Biosystems™ CPG (Controlled Pore Glass) Synthesis Column, 1.0 μmol, 500 Angstrom Pore Size, dG(iBu)
The 1.0μmol dG(iBu) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides<50 bases in length. These columns deliver high-quality oligonucleotides.
Supplier: Applied Biosystems™ 400947
Description
These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.- Proprietary design and manufacturing methods ensure consistency and high quality
- Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
- The 500 angstrom pore size allows synthesis of oligonucleotides of up to 50 bases.
Specifications
Shipped at Room Temperature | |
1 Column |
For Research Use Only. Not for use in diagnostic procedures.