Learn More
Applied Biosystems™ CPG (Controlled Pore Glass) Synthesis Column, 1.0 μmol, 1000 Angstrom Pore Size, dG(iBu)
The 1.0μmol dG(iBu) Controlled Pore Glass (CPG) columns are suitable for the synthesis of any length oligonucleotide. These columns deliver high-quality oligonucleotides.
Supplier: Applied Biosystems™ 401439
Description
These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.- Proprietary design and manufacturing methods ensure consistency and high quality
- Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
- The 1,000 angstrom pore size allows synthesis of oligonucleotides of varying length
Specifications
Shipped at Room Temperature | |
1 Column |
For Research Use Only. Not for use in diagnostic procedures.