Learn More
Therapak™ Courier Shipping Systems
For variable temperature shipments by ground or air
$329.52 - $532.52
Specifications
Applications For specimen transport
Material Urethane
View More Specs
Products 3
Catalog Number Mfr. No. Type Size Price Quantity  
Catalog Number Mfr. No. Type Size Price Quantity  
22-130-209
View Documents
Therapak™
36518
Shipper Medium
Each for $459.12
Only null left
22-130-208
View Documents
Therapak™
36517
Shipper Small
Each for $329.52
Only null left
22-130-211
View Documents
Therapak™
36520
Courier Tote 3 small, 2 medium, 1 large; or 2 small and 1 medium
Each for $532.52
Only null left
Description
Description
  • For sample shipments that require temperature integrity from 24 to 96 hours—ideal for international shipments
  • Reusable urethane shippers with durable, thick wall construction provide more than twice the R-value of EPS shippers
  • Courier tote can be configured to accommodate various configurations, including: three small shippers, two medium shippers, one large shipper, or two small and one medium shipper
  • Replacement lids available separately
Specifications
Specifications
For specimen transport
Urethane
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'