Learn More
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
$702.19 - $1107.94
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
07-200-221
|
Corning™
C3517 |
Black |
Case of 200 Each for $1,107.94
|
|
|||||
07-200-222
|
Corning™
3519 |
White |
Case of 200 Each for $702.19
|
|
|||||
Description
- Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
- Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
- Each carrier kit contains eight carriers and eight handles
Specifications
Tray | |
25/Pk. | |
200/Cs. |