Learn More
Corning™ Sealing Mat for 384-Well Polypropylene Storage Plates
Ideal for use in long-term, low temperature compound storage
Supplier: Corning™ 3341
Description
- Precise fit for 384 well Deep Well Polypropylene Storage Block (Item #’s 3342 and 3347)
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
- Firm seal prevents evaporation and gas exchange
- Ideal for use in long-term, low temperature compound storage
Specifications
DNase and RNase-free | |
Non-sterile | |
50/Cs. | |
Sealing Mat | |
384-well Deep Well Storage Block |
Polypropylene | |
384 | |
10/Pk. | |
V |