missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PET Storage Bottles, Square with Cap
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
$171.36 - $556.50
Specifications
Color | Orange |
---|---|
Diameter (English) Inner Neck | 1.4 in. |
Diameter (Metric) Inner Neck | 37 mm |
For Use With (Application) | Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage |
Material | PET |
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | ||||
07-201-600
|
Corning™
431530 |
125 mL | 50 to 125 mL |
Case of 24 Each for $171.36
|
|
||||
07-201-601
|
Corning™
431531 |
250 mL | 50 to 250 mL |
Case of 24 Each for $261.04
|
|
||||
07-201-602
|
Corning™
431532 |
500 mL | 100 to 500 mL |
Case of 24 Each for $309.25
|
|
||||
07-201-603
|
Corning™
431533 |
1 L | 100 to 1000 mL |
Case of 24 Each for $556.50
|
|
||||
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10â6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic
Specifications
Orange | |
37 mm | |
PET | |
12/Pk. | |
Sterile | |
Orange | |
Single-use | |
Square | |
GL 45 |
1.4 in. | |
Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage | |
24/Cs. | |
12 Pk. | |
GL 45 | |
ISO 9001:2008 | |
Yes | |
HDPE | |
Plug Seal |