missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Low-Volume 384-Well Polystyrene Plates

High-base design is compatible with automation and bar coding

$3179.09 - $3532.75

Specifications

Format Standard
Lid Without Lid
Array 16 x 24
Material Polystyrene
No. of Wells 384
View More Specs

Products 2
Catalog Number Mfr. No. Surface Treatment Price Quantity  
Catalog Number Mfr. No. Surface Treatment Price Quantity  
07-202-625
View Documents
Corning™
C3540
Non-treated
Case of 50 Each for $3,179.09
Only null left
 
09-761-84
View Documents
Corning™
3544
Non-binding
Case of 50 Each for $3,532.75
Only null left
 
Description

Description

  • Validated for the FLIPR™ TETRA system—outer wells aligned for proper viewing with the FLIPR system
  • 2.8mm distance between well bottoms and plate bottom
  • Round wells with flat bottoms
  • Working volume: 5–45μL

  • Untreated surface binds biomolecules through passive interactions; Nonbinding surface (NBS™) creates a nonionic hydrophilic surface that minimizes molecular interactions; Tissue culture-treated surface provides optimal cell attachment

Specifications

Specifications

Standard
16 x 24
384
Non-sterile
For miniaturized 384-well fluorescent homogeneous assays
Without Lid
Polystyrene
0.005 to 0.04 mL
Flat Bottom (F-well)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'