Learn More
Corning™ glutaGRO™ DMEM/F-12 Medium
Used for culture of a variety of cell types under low serum conditions.
Supplier: Corning™ 10103CV
Description
(This is suggested replacement for 10-103-CVR.)
[+] Corning glutagro™ This 1:1 mixture of Dulbecco’s Modified Eagle Medium and Ham’s F-12 media is ideal for culture of a variety of cell types under low serum conditions. Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME. Ham’s F-12 media is based on Ham’s F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids. This formulation of DMEM/Ham’s F-12 is supplemented with Corning glutagro™ in place of L-glutamine for improved stability.
Technical Advantage
Greatly improved stability over the L-glutamine media supplement Dulbecco's Modification of Eagle's Medium/Ham's F-12 50/50 Mix. Corning media certificates are available on our Resource Library.
Specifications
2° to 8°C | |
DMEM/F-12 Medium | |
7.2 |
Liquid | |
Sterile | |
6 x 500 mL |