Learn More
Corning™ BioCoat™ Fibronectin HTS Multiwell Angiogenesis
Provides a standard format to increase the quality and reproducibility of in vitro endothelial cell migration assays
Supplier:  Corning™ 354144
 View more versions of this product
Catalog No. 15100204
Only null left
Explore available promotions
Includes: A Falcon 24- and 96-Multiwell™ insert plate containing a fluorescence blocking, microporous polyethylene terephthalate (PET) membrane evenly coated with human fibronectin, an untreated receiver plate, and a lid
Description
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Specifications
0.3 cm2
3 μm
High Throughput Screening
5/Pk.
Fibronectin
PET
Multiwell Insert Plate
5
5/Cs.
24
Product Suggestions
Product Suggestions