missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Metal, Shot, ACS, 99.9%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C169625KG
Specifications
7440-50-8 | |
Cu | |
2.5 kg | |
[Cu] | |
63.55 | |
ACS |
1 | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
99.9% | |
Poly Bottle |
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
[Cu] |
Cu | |
MFCD00010965 | |
copper |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.