missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) oxide, Honeywell Fluka™
Puriss. p.a.,≥99.0% (RT), powder
$312.08 - $1060.45
Chemical Identifiers
CAS | 1317-38-0 |
---|---|
Molecular Formula | CuO |
Molecular Weight (g/mol) | 79.55 |
MDL Number | MFCD00010979 |
InChI Key | KKCXRELNMOYFLS-UHFFFAOYSA-N |
Synonym | cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide |
PubChem CID | 14829 |
IUPAC Name | oxocopper |
SMILES | [O--].[Cu++] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1317-38-0 | |
79.55 | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
14829 | |
[O--].[Cu++] |
CuO | |
MFCD00010979 | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
oxocopper |
Specifications
1317-38-0 | |
CuO | |
UN3077 | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
oxocopper | |
14829 | |
≥99% (RT) |
100 g | |
MFCD00010979 | |
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide | |
[O--].[Cu++] | |
79.55 | |
79.55g/mol | |
Copper(II) oxide |
Safety and Handling
P273
RTECSNumber : GL7900000