missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, 97%, Honeywell Fluka™

Powder, <5 μm, 97%

$939.70 - $939.70

Chemical Identifiers

CAS 1317-39-1
MDL Number MFCD00010974
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045171
View Documents
Honeywell-Fluka
2088252KG
2 kg Poly Bottle
Each for $939.70
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1317-39-1
MFCD00010974

Specifications

1230°C
1317-39-1
Powder
Cu2O
UN3077
97%
6g/mL at 25°C
Brown or Red
8.0 (at 100g/L 20°C)
2 kg
MFCD00010974
143.09g/mol
Poly Bottle
Copper(I) Oxide
Videos
Safety and Handling

Safety and Handling

P261-P301 + P312 + P330-P305 + P351 + P338

H302 + H332-H319-H410

DOTInformation : 9/PGIII

EINECSNumber : 215-270-7

RTECSNumber : GL8050000

SDS
missing translation for 'documents'

missing translation for 'documents'