missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, Honeywell™

Reagent Grade, 97%

$674.68 - $674.68

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002090
View Documents
Honeywell Chemicals
2129462KG
2 kg
Each for $674.68
Only null left
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7758-89-6
98.996
OXBLHERUFWYNTN-UHFFFAOYSA-M
62652
chlorocopper
ClCu
MFCD00010971
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
CHEBI:53472
Cl[Cu]

Specifications

7758-89-6
ClCu
MFCD00010971
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
OXBLHERUFWYNTN-UHFFFAOYSA-M
chlorocopper
62652
99g/mol
Reagent
Copper(I) Chloride
2 kg
CuCl
2802
Partially soluble in dimethyl sulfide, Insoluble in water
Cl[Cu]
98.996
CHEBI:53472
97%
1.3mmHg (546°C)
Videos
Safety and Handling

Safety and Handling

P273-P280-P305 + P351 + P338-P501

H302-H315-H318-H410

EINECSNumber : 231-842-9

RTECSNumber : GL6990000

SDS
missing translation for 'documents'

missing translation for 'documents'