missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Controlled Environment Coverall
Fisherbrand microporous composite coveralls are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day.
Supplier: Fisherbrand™ CV640322
Description
- Microporous composite material
- Strong and lightweight
- Tapered collar
- Zipper front
- Elastic wrist, ankle and back
- Strong stress-resistant serged seams
Specifications
Cleanroom Coveralls | |
Unisex | |
White | |
Standard | |
Elastic | |
Unhooded | |
Serged | |
Particle | |
Elastic Back | |
None | |
Controlled and Critical Environments | |
Class 100 (ISO 5) |
Medium | |
Microporous Composite Fabric | |
Single-use | |
Elastic | |
Collared | |
Zipper | |
Non-sterile | |
Tapered | |
Yes | |
Set-in Sleeve | |
25/Cs. |