missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Boot Cover

Fisherbrand high top anti-skid boot covers fit over the calf with an elastic top to provide a comfortable fit and to prevent the boot cover from sliding down. Sole material is clean and durable. Upper material is soft and strong for all day comfort.

$492.26 - $567.48

Specifications

Material Polyethylene, Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-165-013
View Documents
Fisherbrand™
BTA1612BFS
Universal
Case of 100 Pair for $492.26
Only null left
 
19-165-013XL
View Documents
Fisherbrand™
BTA1613BFS
X-Large
Case of 100 Pair for $526.46
Only null left
 
19-165-0132X
View Documents
Fisherbrand™
BTA1614BFS
2X-Large
Case of 100 Pair for $567.48
Only null left
 
Description

Description

  • Inherent non-skid physical properties
  • Strong enough to withstand even the most abrasive floor surfaces
  • Fabric provides barrier protection and exhibits low particle generation
  • High COF fit
Specifications

Specifications

Polyethylene, Polypropylene
White
Non-sterile
Anti-skid
Controlled and Critical Environments
Double Bagged
Controlled Environments Boot Covers
Yes
Single-use
Elastic Top and Ankles
Serged
200/Cs.
Abrasion
Class 100 (ISO 5)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'