missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 200 μS/cm at 25°C (95 ppm TDS as NaCl), Ricca Chemical

$118.88 - $395.97

Specifications

Packaging Natural Poly Bottle
Boiling Point 100°C
Type Conductivity Standard
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
2238-16
View Documents
Ricca Chemical Company
223816
500 mL
Each for $118.88
Only null left
 
2238-32
View Documents
Ricca Chemical Company
223832
1 L
Each for $186.53
Only null left
 
2238-1
View Documents
Ricca Chemical Company
22381
4 L
Each for $395.97
Only null left
 
Description

Description

Specifications

Specifications

Natural Poly Bottle
100°C
Conductivity Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'