missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, NIST Traceable, Reagents
Conductivity Standard, 5 μS/cm at 25°C NIST Traceable, Standard, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
$99.47 - $397.90
Specifications
Chemical Name or Material | Conductivity Standard, 5 μS/cm at 25°C NIST Traceable |
---|---|
Packaging | Poly Bottle |
CAS | 7732-18-5 |
Specific Gravity | 1 |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-336-717
|
Labchem Inc
C70000051A |
1 L |
Each for $132.67
|
|
|||||
01-336-716
|
Labchem Inc
C70000054A |
4 L |
Each for $397.90
|
|
|||||
01-336-715
|
Labchem Inc
C7000005500A |
500 mL |
Each for $99.47
|
|
|||||
Specifications
Conductivity Standard, 5 μS/cm at 25°C NIST Traceable | |
7732-18-5 | |
Liquid | |
Calibration of Conductivity Instrument | |
6.7 to 7.3 | |
Conductivity Meter | |
Conductivity Standard |
Poly Bottle | |
1 | |
NaCl/H2O | |
Colorless | |
100°C | |
Completely miscible |