Learn More
OHAUS Compact Scale
With Legal for Trade certifications and a food-safe design that is NSF and HACCP certified, the Valor 1000 series is a reliable choice for general food weighing. Easy-to-operate Valor 1000 scales feature two-button operation, large backlit LCD display and up to 1,500-hour battery life.
Supplier: OHAUS 30554443
Description
- NSF-certified and USDA AMS-accepted design supports HACCP systems, making Valor 1000 perfect for food preparation tasks.
- Stainless steel pans and plastic in-use covers available for extra protection.
- Save money on batteries.
- Energy-efficient design operates for up to one year or up to 1,500 hours.
- Intuitive two-button operation allows for quick weight checking to simplify repetitive operations and increase operator efficiency.
Specifications
5 kg | |
0.5 g | |
Scale |
Backlit LCD | |
V12P5T | |
Class III |