Learn More
OHAUS Compact Scale
With Legal for Trade certifications and a food-safe design that is NSF and HACCP certified, the Valor 1000 series is a reliable choice for general food weighing. Easy-to-operate Valor 1000 scales feature two-button operation, large backlit LCD display and up to 1,500-hour battery life.
Supplier:  OHAUS 30554444
 View more versions of this product
Catalog No. 01-920-224
Only null left
Explore available promotions
Description
Description
  • NSF-certified and USDA AMS-accepted design supports HACCP systems, making Valor 1000 perfect for food preparation tasks.
  • Stainless steel pans and plastic in-use covers available for extra protection.
  • Save money on batteries.
  • Energy-efficient design operates for up to one year or up to 1,500 hours.
  • Intuitive two-button operation allows for quick weight checking to simplify repetitive operations and increase operator efficiency.
Specifications
Specifications
10 kg
1 g
Scale
Backlit LCD
V12P10T
Class III
Product Suggestions
Product Suggestions