missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collodion, USP, Spectrum™ Chemical
CO120, 9004-70-0
$362.14 - $35630.46
Chemical Identifiers
CAS | 9004-70-0 |
---|---|
Molecular Formula | C24H36N8O38 |
Molecular Weight (g/mol) | 1044.57 |
InChI Key | OWWGYSXLINVAES-UHFFFAOYNA-N |
IUPAC Name | 2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
SMILES | OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18601814
|
Spectrum Chemical Mfg Cor
CO120100MLGL |
100 mL | Amber Glass Bottle |
Each for $362.14
|
|
||||
18601815
|
Spectrum Chemical Mfg Cor
CO120500MLGL |
500 mL | Amber Glass Bottle |
Each for $548.06
|
|
||||
18601816
|
Spectrum Chemical Mfg Cor
CO1204LTGL |
4 L | Amber Glass Bottle |
Each for $2,380.57
|
|
||||
18601817
|
Spectrum Chemical Mfg Cor
CO120200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
9004-70-0 | |
1044.57 | |
2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
C24H36N8O38 | |
OWWGYSXLINVAES-UHFFFAOYNA-N | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O |
Specifications
9004-70-0 | |
C24H36N8O38 | |
100 mL | |
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O | |
1044.57 | |
USP |
100% | |
UN2059 | |
OWWGYSXLINVAES-UHFFFAOYNA-N | |
2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol | |
5% | |
Amber Glass Bottle |